Gene Transcripts

Gene Symbol
Rabgef1
Entrez Gene ID
56715
Gene Name
RAB guanine nucleotide exchange factor (GEF) 1
Gene Aliases
Rab5ef, Rabex5, Rin2
Chromosome Location
Chr. 5: 130171791 - 130214342 on Build GRCm38
UniGene ID
Mm.288639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199059.1 NP_001185988.1 6 709
NM_019983.2 NP_064367.1 5 854
GenBank mRNA AF093590.1 - 6 685
AK044568.1 - 5 874
AK146734.1 - 6 659
BC018229.1 - 5 854 4507477

View Full Product Details