Gene Transcripts

Gene Symbol
Chst11
Entrez Gene ID
58250
Gene Name
carbohydrate sulfotransferase 11
Gene Aliases
1110020P09Rik, C4ST, C4ST-1, C4ST1, C4s
Chromosome Location
Chr. 10: 82985380 - 83195907 on Build GRCm38
UniGene ID
Mm.360747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021439.2 NP_067414.2 3 1363
GenBank mRNA AB030378.1 - 3 1515
AJ289133.1 - 3 1371
BC144793.1 - 3 1096 9053341

View Full Product Details