Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
65257
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
2400011J03Rik, Asb-3
Chromosome Location
Chr. 11: 30954194 - 31102704 on Build GRCm38
UniGene ID
Mm.440539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023906.3 NP_076395.2 2 297
GenBank mRNA AF155354.1 - 1 112
AK010315.1 - 2 233
AK028489.1 - 2 290
AK038086.1 - 2 278
AK038897.1 - 2 248
AK143580.1 - 2 232
BB618690.1 - 2 299
BC023086.1 - 2 231 4947610

View Full Product Details