Gene Transcripts
- Gene Symbol
- Brd3
- Entrez Gene ID
- 67382
- Gene Name
- bromodomain containing 3
- Gene Aliases
- 2410084F24Rik, AW060456, Fsrg2, ORFX, RINGL3
- Chromosome Location
- Chr. 2: 27445579 - 27507657 on Build GRCm38
- UniGene ID
- Mm.28721
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113573.1 |
NP_001107045.1
|
5, 6 |
895 |
|
NM_001113574.1 |
NP_001107046.1
|
5, 6 |
895 |
|
NM_023336.4 |
NP_075825.3
|
5, 6 |
1055 |
|
GenBank mRNA |
AB206708.2 |
- |
4, 5 |
706 |
|
AB212272.1 |
- |
4, 5 |
704 |
|
AF269193.1 |
- |
5, 6 |
843 |
|
AK037435.1 |
- |
5, 6 |
877 |
|
AK076472.1 |
- |
5, 6 |
826 |
|
AK076924.1 |
- |
5, 6 |
974 |
|
AK081471.1 |
- |
5, 6 |
876 |
|
AK136007.1 |
- |
5, 6 |
855 |
|
AK136429.1 |
- |
5, 6 |
826 |
|
AK159827.1 |
- |
5, 6 |
881 |
|
AK160752.1 |
- |
5, 6 |
854 |
|
AK161164.1 |
- |
5, 6 |
851 |
|
AK167573.1 |
- |
5, 6 |
853 |
|
AY513269.1 |
- |
4, 5 |
721 |
|
BC004832.1 |
- |
5, 6 |
840 |
|
BC031536.1 |
- |
5, 6 |
869 |
4015879 |
CN530570.1 |
- |
3, 4 |
470 |
|
View Full Product Details