Gene Transcripts

Gene Symbol
Akr1cl
Entrez Gene ID
70861
Gene Name
aldo-keto reductase family 1, member C-like
Gene Aliases
4921521F21Rik, AI451536, AI503553
Chromosome Location
Chr. 1: 65013078 - 65032770 on Build GRCm38
UniGene ID
Mm.18725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027582.3 NP_081858.2 9 1105
GenBank mRNA AK014937.1 - 8 984
AK135492.1 - 9 1108
BC051128.1 - 9 1105 6707386

View Full Product Details