Gene Transcripts

Gene Symbol
MUC4
Entrez Gene ID
4585
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP, HSA276359, MUC-4
Chromosome Location
Chr. 3: 195746765 - 195811973 on Build GRCh38
UniGene ID
Hs.369646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322468.1 NP_001309397.1 21058
NM_004532.5 NP_004523.3 16 2332
NM_018406.6 NP_060876.5 15040
NM_138297.4 NP_612154.2 15 2179
GenBank mRNA AJ010901.1 - 16 2116
AJ242541.1 - 17 2341
AJ242542.1 - 15 2041
AJ242543.1 - 16 2040
AJ242544.1 - 16 2012
AJ242545.1 - 15 1951
AJ242546.1 - 14 1686
AJ242547.1 - 16 2116
AJ242548.1 - 12 1438
AJ242549.1 - 16 2216
AJ242550.1 - 15 2063
AJ276359.1 - 18 5226
AJ277412.1 - 17 5070
AJ277505.1 - 16 4967
AJ400633.1 - 18 5240
AJ400849.1 - 18 5150
AJ400850.1 - 18 5150
AJ400851.1 - 18 5240
AJ400852.1 - 17 4994
AJ400853.1 - 17 4966
AJ400854.1 - 17 5071
AJ400855.1 - 17 5042
AJ400856.1 - 18 5197
AJ400857.1 - 18 5226
AJ400858.1 - 18 5226

View Full Product Details