Gene Transcripts

Gene Symbol
MUC4
Entrez Gene ID
4585
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP, HSA276359, MUC-4
Chromosome Location
Chr. 3: 195746765 - 195811973 on Build GRCh38
UniGene ID
Hs.369646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322468.1 NP_001309397.1 19469
NM_004532.5 NP_004523.3 5 743
NM_018406.6 NP_060876.5 13451
NM_138297.4 NP_612154.2 4 590
GenBank mRNA AJ010901.1 - 5 527
AJ242541.1 - 6 752
AJ242542.1 - 5 541
AJ242543.1 - 5 451
AJ242544.1 - 5 451
AJ242545.1 - 5 451
AJ242546.1 - 5 527
AJ242547.1 - 5 527
AJ242548.1 - 5 527
AJ242549.1 - 5 627
AJ242550.1 - 4 474
AJ276359.1 - 7 3637
AJ400633.1 - 7 3651
AJ400849.1 - 7 3561
AJ400850.1 - 7 3561
AJ400851.1 - 7 3651
AJ400856.1 - 7 3637
AJ400857.1 - 7 3637
AJ400858.1 - 7 3637
BX957139.1 - 4 498
DB227644.1 - 3 337
EF091824.1 - 8 3512

View Full Product Details