Gene Transcripts

Gene Symbol
MYBPC2
Entrez Gene ID
4606
Gene Name
myosin binding protein C, fast type
Gene Aliases
MYBPC, MYBPCF
Chromosome Location
Chr. 19: 50432903 - 50466326 on Build GRCh38
UniGene ID
Hs.85937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004533.3 NP_004524.3 23, 24 2836
GenBank mRNA BC130536.1 - 23, 24 2803
BC136389.1 - 23, 24 2803 9020376
X73113.1 - 23, 24 2807

View Full Product Details