Gene Transcripts

Gene Symbol
Setmar
Entrez Gene ID
74729
Gene Name
SET domain without mariner transposase fusion
Gene Aliases
5830404F24Rik, Etet2
Chromosome Location
Chr. 6: 108065045 - 108077127 on Build GRCm38
UniGene ID
Mm.56539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276356.1 NP_001263285.1 3 488
NM_178391.4 NP_848478.2 2 335
GenBank mRNA AK017895.1 - 2 335
BB662956.1 - 2 336

View Full Product Details