Gene Transcripts

Gene Symbol
Flywch2
Entrez Gene ID
76917
Gene Name
FLYWCH family member 2
Gene Aliases
2810417J12Rik, 3010014L17Rik, AW548203
Chromosome Location
Chr. 17: 23776918 - 23786077 on Build GRCm38
UniGene ID
Mm.269613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029798.3 NP_084074.1 2, 3 513
GenBank mRNA AK013108.1 - 2, 3 515
AK019378.1 - 2, 3 501
BC024947.1 - 2, 3 469 2648415

View Full Product Details