Gene Transcripts

Gene Symbol
Rdh1
Entrez Gene ID
107605
Gene Name
retinol dehydrogenase 1 (all trans)
Gene Aliases
Chromosome Location
Chr. 10: 127759726 - 127768299 on Build GRCm38
UniGene ID
Mm.484271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080436.3 NP_536684.2 3 899
GenBank mRNA AK136574.1 - 3 942
AY028928.1 - 3 853
BC137930.1 - 3 817 8860949

View Full Product Details