Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008104.2 | NP_032130.2 | 2, 3 | 370 | |
GenBank mRNA | AF081556.1 | - | 2, 3 | 336 | |
AK135689.1 | - | 3, 4 | 476 | ||
BC110631.1 | - | 2, 3 | 400 | 40054293 | |
BC110632.1 | - | 2, 3 | 400 | 40054295 | |
D88611.1 | - | 2, 3 | 577 |