Gene Transcripts

Gene Symbol
Prm3
Entrez Gene ID
19120
Gene Name
protamine 3
Gene Aliases
BC061127, PX, Pxg
Chromosome Location
Chr. 16: 10790508 - 10790914 on Build GRCm38
UniGene ID
Mm.291887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013638.2 NP_038666.2 1 145
GenBank mRNA AK132894.1 - 1 146
BC049598.1 - 136
BC061127.1 - 144 6771130
BC090986.1 - 205 6740459

View Full Product Details