Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008023.1 | NP_032049.1 | 1 | 308 | |
GenBank mRNA | BC115976.1 | - | 1 | 188 | |
BC139476.1 | - | 1 | 283 | 8862498 | |
BC139477.1 | - | 1 | 283 | 8862499 | |
X92591.1 | - | 1 | 308 |