Gene Transcripts

Gene Symbol
B4galnt4
Entrez Gene ID
330671
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 4
Gene Aliases
BC038881, Gm1095, beta4GalNAcT4
Chromosome Location
Chr. 7: 141061109 - 141072119 on Build GRCm38
UniGene ID
Mm.133741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177897.3 NP_808565.2 17 2642
GenBank mRNA AB114827.1 - 17 2642
BC038881.1 - 3 589
BC062653.1 - 13 2332
BC079533.1 - 6 1545

View Full Product Details