Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014602.2 | NP_055417.1 | 3 | 1352 | |
GenBank mRNA | AK314219.1 | - | 3 | 1299 | |
BC017332.2 | - | 3 | 1317 | ||
BC057239.1 | - | 3 | 1333 | ||
BC110318.1 | - | 3 | 1298 | 5492292 | |
BC127105.1 | - | 3 | 1169 | 40124707 | |
BC127106.1 | - | 3 | 1169 | 40124708 | |
Y08991.1 | - | 3 | 1338 |