Gene Transcripts

Gene Symbol
C1S
Entrez Gene ID
716
Gene Name
complement component 1, s subcomponent
Gene Aliases
Chromosome Location
Chr. 12: 7060676 - 7071032 on Build GRCh38
UniGene ID
Hs.458355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001734.3 NP_001725.1 3 423
NM_201442.2 NP_958850.1 3 510
XM_005253760.1 XP_005253817.1 3 402
GenBank mRNA AK290295.1 - 3 457
BC056903.1 - 3 450 6199186
CD358820.1 - 3 382
DB257339.1 - 3 423
DB265957.1 - 3 510
J04080.1 - 3 349
M18767.1 - 3 369

View Full Product Details