Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173800.4 | NP_776161.3 | 18 | 2851 | |
GenBank mRNA | AK075131.1 | - | 13 | 1386 | |
AK291655.1 | - | 18 | 2865 | ||
AY560010.1 | - | 18 | 2739 | ||
BC036440.1 | - | 13 | 1730 | ||
BC045809.1 | - | 12 | 1448 | ||
BC060869.1 | - | 19 | 2778 | 30346049 | |
BC068560.1 | - | 17 | 2697 | 30341970 | |
BC070028.1 | - | 13 | 1848 | ||
BC094716.1 | - | 13 | 1848 | ||
BC109022.1 | - | 18 | 2715 | 40008501 | |
BC109023.1 | - | 18 | 2715 | 40008502 |