Gene Transcripts

Gene Symbol
USP6
Entrez Gene ID
9098
Gene Name
ubiquitin specific peptidase 6
Gene Aliases
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location
Chr. 17: 5116438 - 5175032 on Build GRCh38
UniGene ID
Hs.448851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304284.1 NP_001291213.1 13, 14 2502
NM_004505.3 NP_004496.2 5, 6 1971
XM_011524050.1 XP_011522352.1 9, 10 2720
XM_011524051.2 XP_011522353.1 11, 12 1815
XM_011524052.2 XP_011522354.1 10, 11 1929
XM_011524053.2 XP_011522355.1 11, 12 1842
XM_011524054.2 XP_011522356.1 11, 12 1809
XM_011524055.2 XP_011522357.1 9, 10 1736
XM_011524056.2 XP_011522358.1 6, 7 1689
XM_011524057.2 XP_011522359.1 10, 11 1918
XM_011524058.2 XP_011522360.1 10, 11 1918
XM_011524059.2 XP_011522361.1 10, 11 1918
XM_017025290.1 XP_016880779.1 8, 9 2558
GenBank mRNA AB449915.1 - 4, 5 272
AY143550.1 - 5, 6 1971
BX647719.1 - 13, 14 2503
X63546.1 - 5, 6 1968
X63547.1 - 5, 6 1968

View Full Product Details