Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002096.2 | NP_002087.2 | 7 | 1103 | |
GenBank mRNA | AK293124.1 | - | 7 | 703 | |
AK302178.1 | - | 4 | 566 | ||
AK315240.1 | - | 7 | 987 | ||
BC000120.1 | - | 7 | 965 | 3352627 | |
BT007097.1 | - | 6 | 818 | ||
X64002.1 | - | 7 | 881 | ||
X64037.1 | - | 7 | 996 |