Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193493.1 | NP_001180422.1 | 7 | 1217 | |
NM_003038.4 | NP_003029.2 | 8 | 2085 | ||
XM_006712079.2 | XP_006712142.1 | 8 | 1451 | ||
GenBank mRNA | AK126636.1 | - | 7 | 3275 | |
AK315986.1 | - | 9 | 2007 | ||
BC026216.1 | - | 8 | 1986 | 4420405 | |
BC072423.1 | - | 8 | 2045 | 6186177 | |
L14595.1 | - | 8 | 1984 |