Gene Transcripts
- Gene Symbol
- SLC22A14
- Entrez Gene ID
- 9389
- Gene Name
- solute carrier family 22 member 14
- Gene Aliases
- OCTL2, OCTL4, ORCTL4
- Chromosome Location
- Chr. 3: 38281662 - 38318575 on Build GRCh38
- UniGene ID
- Hs.165559
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320033.1 |
NP_001306962.1
|
10, 11 |
1787 |
|
NM_004803.4 |
NP_004794.2
|
9, 10 |
1832 |
|
XM_005265585.4 |
XP_005265642.1
|
10, 11 |
2609 |
|
XM_006713416.3 |
XP_006713479.2
|
10, 11 |
2533 |
|
XM_006713417.3 |
XP_006713480.2
|
10, 11 |
2500 |
|
XM_006713418.3 |
XP_006713481.2
|
10, 11 |
2420 |
|
XM_011534245.2 |
XP_011532547.1
|
10, 11 |
2621 |
|
GenBank mRNA |
AB011082.1 |
- |
9, 10 |
1831 |
|
AK315317.1 |
- |
10, 11 |
1785 |
|
BC075070.2 |
- |
9, 10 |
1776 |
30915378 |
BC075071.2 |
- |
9, 10 |
1776 |
30915591 |
BC126482.1 |
- |
9, 10 |
1813 |
8992198 |
BC130543.1 |
- |
9, 10 |
1813 |
40146574 |
BC144079.1 |
- |
9, 10 |
1772 |
|
View Full Product Details