Gene Transcripts

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
Chromosome Location
Chr. 15: 74541177 - 74598131 on Build GRCh38
UniGene ID
Hs.647642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307939.1 NP_001294868.1 9 3316
NM_006465.3 NP_006456.1 9 3313
GenBank mRNA AB208830.1 - 8 3360
AF116846.1 - 9 3244
AL137417.1 - 1 338
BC045545.1 - 9 3287

View Full Product Details