Gene Transcripts

Gene Symbol
SLC22A15
Entrez Gene ID
55356
Gene Name
solute carrier family 22 member 15
Gene Aliases
FLIPT1, PRO34686
Chromosome Location
Chr. 1: 115976498 - 116070054 on Build GRCh38
UniGene ID
Hs.125482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018420.2 NP_060890.2 12 1842
GenBank mRNA AL353933.1 - 4 543
BC026358.1 - 12 1841 4814675

View Full Product Details