Gene Transcripts

Gene Symbol
SLC4A9
Entrez Gene ID
83697
Gene Name
solute carrier family 4 member 9
Gene Aliases
AE4
Chromosome Location
Chr. 5: 140359645 - 140375177 on Build GRCh38
UniGene ID
Hs.550313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258426.1 NP_001245355.1 6 802
NM_001258427.1 NP_001245356.1 6 802
NM_001258428.1 NP_001245357.1 6 874
NM_031467.2 NP_113655.2 6 802
XM_005268521.3 XP_005268578.2 6 1359
XM_011537694.2 XP_011535996.1 6 1431
XM_017009933.1 XP_016865422.1 6 1431
XM_017009934.1 XP_016865423.1 6 1431
XM_017009935.1 XP_016865424.1 6 1431
XM_017009936.1 XP_016865425.1 6 1359
XM_017009937.1 XP_016865426.1 5 1317
XM_017009938.1 XP_016865427.1 6 1431
XM_017009939.1 XP_016865428.1 6 1359
XM_017009940.1 XP_016865429.1 6 1431
XM_017009941.1 XP_016865430.1 6 1431
XM_017009942.1 XP_016865431.1 6 1431
XM_017009943.1 XP_016865432.1 5 742
GenBank mRNA AB032762.1 - 6 761
AF313465.1 - 6 861
AF332961.1 - 6 802
AF336237.1 - 6 794
BC136262.1 - 6 793 9020249
BC143602.1 - 6 793 9052115

View Full Product Details