Gene Transcripts

Gene Symbol
C22orf23
Entrez Gene ID
84645
Gene Name
chromosome 22 open reading frame 23
Gene Aliases
EVG1, dJ1039K5.6
Chromosome Location
Chr. 22: 37943050 - 37953669 on Build GRCh38
UniGene ID
Hs.517612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207062.1 NP_001193991.1 6 806
NM_032561.4 NP_115950.3 7 1017
XM_005261781.1 XP_005261838.1 6 1151
XM_005261782.3 XP_005261839.1 6 1031
XM_005261783.2 XP_005261840.1 5 1088
XM_005261784.2 XP_005261841.1 6 918
GenBank mRNA AF324466.1 - 6 817
AK057349.1 - 7 1067
BC031998.1 - 7 899 4755328
CR456440.1 - 7 897

View Full Product Details