Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206925.1 | NP_001193854.1 | 2 | 395 | |
NM_006889.4 | NP_008820.3 | 3 | 500 | ||
NM_175862.4 | NP_787058.4 | 3 | 445 | ||
NM_176892.1 | NP_795711.1 | 3 | 500 | ||
GenBank mRNA | AK291497.1 | - | 3 | 353 | |
AK301237.1 | - | 2 | 391 | ||
AK313103.1 | - | 3 | 354 | ||
AK316203.1 | - | 2 | 391 | ||
BC040261.1 | - | 3 | 433 | 5173789 | |
CD705810.1 | - | 3 | 522 | ||
CR541844.1 | - | 2 | 299 | ||
DA619360.1 | - | 3 | 446 | ||
L25259.1 | - | 3 | 423 | ||
U04343.1 | - | 3 | 446 |