Gene Transcripts

Gene Symbol
GYPA
Entrez Gene ID
2993
Gene Name
glycophorin A (MNS blood group)
Gene Aliases
CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2
Chromosome Location
Chr. 4: 144109303 - 144140854 on Build GRCh38
UniGene ID
Hs.434973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308187.1 NP_001295116.1 6 1850
NM_001308190.1 NP_001295119.1 6 1790
NM_002099.7 NP_002090.4 7 1889
XM_017008134.1 XP_016863623.1 6 1814
XM_017008135.1 XP_016863624.1 8 1956
XM_017008136.1 XP_016863625.1 5 1779
GenBank mRNA BX649043.1 - 1 1761
L31860.1 - 7 1782
X51798.1 - 7 1755

View Full Product Details