Gene Transcripts

Gene Symbol
SIRPD
Entrez Gene ID
128646
Gene Name
signal regulatory protein delta
Gene Aliases
PTPNS1L2, dJ576H24.4
Chromosome Location
Chr. 20: 1534251 - 1557697 on Build GRCh38
UniGene ID
Hs.729560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178460.2 NP_848555.2 2 257
GenBank mRNA AK093083.1 - 2 1403
BC033502.1 - 2 257 5172531
DA466740.1 - 3 457

View Full Product Details