Gene Transcripts
- Gene Symbol
- UGGT2
- Entrez Gene ID
- 55757
- Gene Name
- UDP-glucose glycoprotein glucosyltransferase 2
- Gene Aliases
- HUGT2, UGCGL2, UGT2
- Chromosome Location
- Chr. 13: 95801580 - 96053482 on Build GRCh38
- UniGene ID
- Hs.193226
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020121.3 |
NP_064506.3
|
19, 20 |
2377 |
|
XM_011521094.1 |
XP_011519396.1
|
19, 20 |
2332 |
|
XM_011521095.1 |
XP_011519397.1
|
19, 20 |
2332 |
|
XM_011521096.1 |
XP_011519398.1
|
19, 20 |
2332 |
|
XM_011521097.1 |
XP_011519399.1
|
19, 20 |
2332 |
|
XM_011521098.1 |
XP_011519400.1
|
12, 13 |
1349 |
|
XM_011521099.2 |
XP_011519401.1
|
11, 12 |
1266 |
|
XM_011521100.2 |
XP_011519402.1
|
19, 20 |
2332 |
|
XM_011521102.2 |
XP_011519404.1
|
9, 10 |
1040 |
|
XM_011521103.2 |
XP_011519405.1
|
9, 10 |
1098 |
|
XM_017020662.1 |
XP_016876151.1
|
19, 20 |
2332 |
|
XM_017020663.1 |
XP_016876152.1
|
20, 21 |
2315 |
|
GenBank mRNA |
AF227906.2 |
- |
19, 20 |
2278 |
|
AK001735.1 |
- |
19, 20 |
2302 |
|
AL133051.1 |
- |
16, 17 |
1757 |
|
BC125233.1 |
- |
19, 20 |
2233 |
40124676 |
View Full Product Details