Gene Transcripts

Gene Symbol
SQRDL
Entrez Gene ID
58472
Gene Name
sulfide quinone reductase-like (yeast)
Gene Aliases
CGI-44, PRO1975, SQOR
Chromosome Location
Chr. 15: 45631148 - 45691294 on Build GRCh38
UniGene ID
Hs.511251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271213.1 NP_001258142.1 7 873
NM_021199.3 NP_067022.1 6 1066
GenBank mRNA AF042284.1 - 6 756
AF118085.1 - 5 587
AF151802.1 - 6 750
AK130140.1 - 7 873
AK222463.1 - 6 765
AK313111.1 - 6 745
BC016836.1 - 6 721 3884024

View Full Product Details