Gene Transcripts

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
Chromosome Location
Chr. 9: 72900662 - 72953317 on Build GRCh38
UniGene ID
Hs.76392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000689.4 NP_000680.2 6 879
GenBank mRNA AB209821.1 - 6 605
AF003341.1 - 6 562
AK000118.1 - 6 606
AK026641.1 - 6 606
AK293176.1 - 6 572
AY390731.1 - 6 562
BC001505.2 - 6 587 2988388
BG773210.1 - 6 621
BT006921.1 - 6 562
EU668318.1 - 6 606
EU794589.1 - 6 562
FJ224286.1 - 6 615
K03000.1 - 2 79
M26761.1 - 2 79

View Full Product Details