Gene Transcripts
- Gene Symbol
- AGBL4
- Entrez Gene ID
- 84871
- Gene Name
- ATP/GTP binding protein like 4
- Gene Aliases
- CCP6
- Chromosome Location
- Chr. 1: 48520157 - 50024011 on Build GRCh38
- UniGene ID
- Hs.551844
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001323573.1 |
NP_001310502.1
|
6, 7 |
822 |
|
NM_001323574.1 |
NP_001310503.1
|
6, 7 |
822 |
|
NM_001323575.1 |
NP_001310504.1
|
6, 7 |
786 |
|
NM_032785.3 |
NP_116174.3
|
6, 7 |
786 |
|
XM_011542308.2 |
XP_011540610.1
|
6, 7 |
806 |
|
XM_017002595.1 |
XP_016858084.1
|
6, 7 |
843 |
|
XM_017002596.1 |
XP_016858085.1
|
6, 7 |
843 |
|
XM_017002597.1 |
XP_016858086.1
|
6, 7 |
843 |
|
GenBank mRNA |
AK027348.1 |
- |
2, 3 |
165 |
|
AK094826.1 |
- |
6, 7 |
745 |
|
AK294394.1 |
- |
6, 7 |
781 |
|
AK296221.1 |
- |
6, 7 |
786 |
|
BC126383.1 |
- |
2, 3 |
98 |
8992099 |
View Full Product Details