Gene Transcripts

Gene Symbol
CD5
Entrez Gene ID
921
Gene Name
CD5 molecule
Gene Aliases
LEU1, T1
Chromosome Location
Chr. 11: 61102458 - 61127852 on Build GRCh38
UniGene ID
Hs.58685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014207.3 NP_055022.2 11 2357
GenBank mRNA BC027901.1 - 11 2320

View Full Product Details