Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030783.1 | NP_110410.1 | 2, 3 | 462 | |
XM_005253153.1 | XP_005253210.1 | 2, 3 | 395 | ||
XM_011520391.2 | XP_011518693.1 | 2, 3 | 1032 | ||
XM_011520392.2 | XP_011518694.1 | 3, 4 | 459 | ||
XM_017018368.1 | XP_016873857.1 | 4, 5 | 2854 | ||
XM_017018369.1 | XP_016873858.1 | 3, 4 | 584 | ||
GenBank mRNA | AK311723.1 | - | 2, 3 | 394 | |
AL834357.1 | - | 2, 3 | 376 | ||
BC001210.1 | - | 2, 3 | 462 | 3503123 |