Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243146.1 | NP_001230075.1 | 10 | 1020 | |
NM_001243147.1 | NP_001230076.1 | 11 | 1071 | ||
NM_002533.3 | NP_002524.2 | 12 | 1344 | ||
NM_206840.2 | NP_996671.1 | 11 | 1270 | ||
XM_011544196.1 | XP_011542498.1 | 13 | 1408 | ||
XM_011544198.1 | XP_011542500.1 | 12 | 1363 | ||
XM_011544199.1 | XP_011542501.1 | 12 | 1317 | ||
XM_011544200.1 | XP_011542502.1 | 11 | 1291 | ||
XM_017001378.1 | XP_016856867.1 | 14 | 1517 | ||
XM_017001379.1 | XP_016856868.1 | 13 | 1436 | ||
XM_017001380.1 | XP_016856869.1 | 14 | 1517 | ||
XM_017001381.1 | XP_016856870.1 | 14 | 1514 | ||
XM_017001382.1 | XP_016856871.1 | 13 | 1447 | ||
XM_017001383.1 | XP_016856872.1 | 12 | 1366 | ||
XM_017001384.1 | XP_016856873.1 | 14 | 1517 | ||
XM_017001385.1 | XP_016856874.1 | 14 | 1517 | ||
XM_017001386.1 | XP_016856875.1 | 10 | 1210 | ||
GenBank mRNA | AK297070.1 | - | 12 | 1303 | |
AK297396.1 | - | 10 | 971 | ||
AK298244.1 | - | 11 | 1020 | ||
AK298826.1 | - | 11 | 1222 | ||
BC012105.1 | - | 10 | 931 | 4636004 | |
U68140.1 | - | 12 | 1338 | ||
U78772.1 | - | 11 | 1251 |