Gene Transcripts
- Gene Symbol
- ACOT9
- Entrez Gene ID
- 23597
- Gene Name
- acyl-CoA thioesterase 9
- Gene Aliases
- ACATE2, CGI-16, MT-ACT48, MTACT48
- Chromosome Location
- Chr. X: 23701055 - 23743290 on Build GRCh38
- UniGene ID
- Hs.298885
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033583.2 |
NP_001028755.2
|
5, 6 |
519 |
|
NM_001037171.1 |
NP_001032248.1
|
6, 7 |
546 |
|
XM_005274472.2 |
XP_005274529.1
|
4, 5 |
418 |
|
GenBank mRNA |
AF132950.1 |
- |
4, 5 |
421 |
|
AK000659.1 |
- |
5, 6 |
505 |
|
AK024337.1 |
- |
7, 8 |
1611 |
|
AL548456.3 |
- |
6, 7 |
546 |
|
BC012573.1 |
- |
5, 6 |
490 |
|
BC136671.1 |
- |
5, 6 |
470 |
9020661 |
BC144360.1 |
- |
5, 6 |
470 |
9052881 |
BC144370.1 |
- |
5, 6 |
470 |
|
BX648512.1 |
- |
6, 7 |
530 |
|
View Full Product Details