Gene Transcripts

Gene Symbol
SLC22A13
Entrez Gene ID
9390
Gene Name
solute carrier family 22 member 13
Gene Aliases
OAT10, OCTL1, OCTL3, ORCTL-3, ORCTL3
Chromosome Location
Chr. 3: 38265807 - 38280030 on Build GRCh38
UniGene ID
Hs.225941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004256.3 NP_004247.2 5 917
XM_017007519.1 XP_016863008.1 4 663
GenBank mRNA AB010438.1 - 5 884
AK298375.1 - 4 663
AK315301.1 - 5 912
BC035973.1 - 5 912 4609915

View Full Product Details