Gene Transcripts

Gene Symbol
SNUPN
Entrez Gene ID
10073
Gene Name
snurportin 1
Gene Aliases
KPNBL, RNUT1, Snurportin1
Chromosome Location
Chr. 15: 75598083 - 75626378 on Build GRCh38
UniGene ID
Hs.21577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042581.1 NP_001036046.1 3 528
NM_001042588.1 NP_001036053.1 3 333
NM_005701.3 NP_005692.1 3 633
GenBank mRNA AF039029.1 - 3 278
AK054641.1 - 4 766
AK289475.1 - 3 329
BC004203.2 - 3 264 3605328
BU945130.1 - 3 427
CD244425.1 - 3 528
CR456811.1 - 2 188

View Full Product Details