Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143838.2 | NP_001137310.1 | 5 | 704 | |
NM_001284509.1 | NP_001271438.1 | 5 | 653 | ||
NM_001284510.1 | NP_001271439.1 | 4 | 575 | ||
NM_177550.4 | NP_808218.1 | 5 | 704 | ||
XM_011523795.2 | XP_011522097.1 | 5 | 694 | ||
GenBank mRNA | AJ489980.1 | - | 4 | 487 | |
AK127797.1 | - | 5 | 601 | ||
AK131206.1 | - | 2 | 1748 | ||
AK172785.1 | - | 5 | 649 | ||
AK297612.1 | - | 4 | 520 | ||
AY151833.1 | - | 5 | 628 | ||
BC104795.1 | - | 5 | 660 | 8143798 | |
BC112151.1 | - | 5 | 660 | 8327619 | |
BC143689.1 | - | 5 | 660 | 9052203 |