Gene Transcripts
- Gene Symbol
- ACSL3
- Entrez Gene ID
- 2181
- Gene Name
- acyl-CoA synthetase long-chain family member 3
- Gene Aliases
- ACS3, FACL3, PRO2194
- Chromosome Location
- Chr. 2: 222860984 - 222944624 on Build GRCh38
- UniGene ID
- Hs.655772
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004457.3 |
NP_004448.2
|
13, 14 |
1999 |
|
NM_203372.1 |
NP_976251.1
|
12, 13 |
1892 |
|
XM_017003584.1 |
XP_016859073.1
|
11, 12 |
1811 |
|
GenBank mRNA |
AB061712.1 |
- |
13, 14 |
1967 |
|
AF116690.1 |
- |
1, 2 |
89 |
|
AK001471.1 |
- |
8, 9 |
918 |
|
AK023191.1 |
- |
10, 11 |
1544 |
|
AK314621.1 |
- |
13, 14 |
1977 |
|
BC041692.1 |
- |
13, 14 |
1991 |
5226905 |
D89053.1 |
- |
11, 12 |
1690 |
|
View Full Product Details