Gene Transcripts

Gene Symbol
BPIFB1
Entrez Gene ID
92747
Gene Name
BPI fold containing family B member 1
Gene Aliases
C20orf114, LPLUNC1
Chromosome Location
Chr. 20: 33283135 - 33309878 on Build GRCh38
UniGene ID
Hs.65551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033197.2 NP_149974.2 12 1317
GenBank mRNA AF364078.1 - 12 1242
AK127971.1 - 9 816
AK290243.1 - 12 1299
AK313176.1 - 12 1235
AY358595.1 - 12 1229
BC008429.1 - 12 1232 4291561

View Full Product Details