Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000038.5 | NP_000029.2 | 4 | 457 | |
NM_001127510.2 | NP_001120982.1 | 5 | 565 | ||
NM_001127511.2 | NP_001120983.2 | 3 | 615 | ||
GenBank mRNA | AB210001.1 | - | 5 | 529 | |
AK289567.1 | - | 4 | 519 | ||
AK294544.1 | - | 3 | 599 | ||
BC056268.1 | - | 4 | 428 | ||
DA795663.1 | - | 4 | 457 | ||
M73548.1 | - | 3 | 410 | ||
M74088.1 | - | 3 | 390 |