Gene Transcripts
- Gene Symbol
- AKR7A2
- Entrez Gene ID
- 8574
- Gene Name
- aldo-keto reductase family 7 member A2
- Gene Aliases
- AFAR, AFAR1, AFB1-AR1, AKR7
- Chromosome Location
- Chr. 1: 19302708 - 19312146 on Build GRCh38
- UniGene ID
- Hs.571886
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320979.1 |
NP_001307908.1
|
4, 5 |
699 |
|
NM_003689.3 |
NP_003680.2
|
5, 6 |
804 |
|
GenBank mRNA |
AF026947.1 |
- |
5, 6 |
772 |
|
BC004111.2 |
- |
5, 6 |
731 |
3832140 |
BC007352.2 |
- |
5, 6 |
779 |
|
BC010852.1 |
- |
5, 6 |
780 |
3898226 |
BC011586.1 |
- |
5, 6 |
731 |
4620484 |
BC012171.1 |
- |
5, 6 |
779 |
4642811 |
BC013996.2 |
- |
5, 6 |
735 |
3625337 |
BT007347.1 |
- |
5, 6 |
695 |
|
BX401279.2 |
- |
4, 5 |
647 |
|
EU794591.1 |
- |
5, 6 |
804 |
|
Y16675.1 |
- |
5, 6 |
766 |
|
View Full Product Details