Gene Transcripts

Gene Symbol
NELFE
Entrez Gene ID
7936
Gene Name
negative elongation factor complex member E
Gene Aliases
D6S45, NELF-E, RD, RDBP, RDP
Chromosome Location
Chr. 6: 31952087 - 31959087 on Build GRCh38
UniGene ID
Hs.423935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002904.5 NP_002895.3 11 1497
GenBank mRNA BU935034.1 - 1 254

View Full Product Details