Gene Transcripts
- Gene Symbol
- UTP11
- Entrez Gene ID
- 51118
- Gene Name
- UTP11, small subunit processome component homolog (S. cerevisiae)
- Gene Aliases
- CGI-94, CGI94, UTP11L
- Chromosome Location
- Chr. 1: 38012712 - 38024825 on Build GRCh38
- UniGene ID
- Hs.472038
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016037.3 |
NP_057121.2
|
4, 5 |
436 |
|
XM_011541555.2 |
XP_011539857.1
|
3, 4 |
370 |
|
GenBank mRNA |
AF067802.1 |
- |
4, 5 |
399 |
|
AF151852.1 |
- |
4, 5 |
414 |
|
AK023587.1 |
- |
2, 3 |
1305 |
|
AK291900.1 |
- |
4, 5 |
366 |
|
AK296018.1 |
- |
4, 5 |
369 |
|
AK308377.1 |
- |
3, 4 |
371 |
|
BC005182.1 |
- |
4, 5 |
381 |
3995984 |
BU940926.1 |
- |
4, 5 |
439 |
|
View Full Product Details