Gene Transcripts

Gene Symbol
CCDC28B
Entrez Gene ID
79140
Gene Name
coiled-coil domain containing 28B
Gene Aliases
Chromosome Location
Chr. 1: 32200386 - 32205390 on Build GRCh38
UniGene ID
Hs.534482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301011.1 NP_001287940.1 5 976
XM_011542113.2 XP_011540415.1 5 1735

View Full Product Details