Gene Transcripts

Gene Symbol
NBPF1
Entrez Gene ID
55672
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
AB13, AB14, AB23, AD2, NBG, NBPF
Chromosome Location
Chr. 1: 16562427 - 16613605 on Build GRCh38
UniGene ID
Hs.467587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017940.4 NP_060410.2 5827
GenBank mRNA AB051480.1 - 21 3884
AF379611.1 - 1 821
AF379612.1 - 1 821
AF379613.1 - 3 1616
AF379614.1 - 1 323
AF379618.1 - 1 589
AF379621.1 - 1 245
AF379624.1 - 23 4083
AF379627.1 - 2 1467
AF379628.1 - 3 1754
BC034418.1 - 16 3389

Gene Symbol
LOC102724250
Entrez Gene ID
102724250
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
Chromosome Location
Chr. 1|NT_187366: 4611 - 41174 on Build GRCh38
UniGene ID
Hs.650616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011546188.2 XP_011544490.1 22 12924

Gene Symbol
NBPF10
Entrez Gene ID
100132406
Gene Name
neuroblastoma breakpoint family member 10
Gene Aliases
AB6, AG1, NBPF9
Chromosome Location
Chr. 1: 146066318 - 146144804 on Build GRCh38
UniGene ID
Hs.714127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF379616.1 - 9 2590

Gene Symbol
NBPF3
Entrez Gene ID
84224
Gene Name
neuroblastoma breakpoint family member 3
Gene Aliases
AE2
Chromosome Location
Chr. 1: 21436775 - 21484900 on Build GRCh38
UniGene ID
Hs.325422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256416.2 NP_001243345.1 15 3439
NM_001256417.2 NP_001243346.1 14 3265
NM_032264.4 NP_115640.1 15 3475
NR_046176.2 18 4004
XM_006710957.1 XP_006711020.1 15 3333
XM_006710958.1 XP_006711021.1 18 3789
XM_006710959.2 XP_006711022.1 16 3573
XM_011542276.1 XP_011540578.1 19 4046
XM_011542277.1 XP_011540579.1 19 4031
XM_011542278.1 XP_011540580.1 19 3860
XM_011542281.1 XP_011540583.1 16 3396
XM_017002498.1 XP_016857987.1 19 3845
XM_017002499.1 XP_016857988.1 14 3111
XM_017002500.1 XP_016857989.1 18 3864
XM_017002501.1 XP_016857990.1 19 3749
XM_017002502.1 XP_016857991.1 18 3678
GenBank mRNA AK095602.1 - 18 3958
AK098438.1 - 5 1720
AL136890.1 - 15 3252
AL834330.1 - 11 2448
BC024011.2 - 15 3366 4827023

View Full Product Details