Gene Transcripts

Gene Symbol
KIAA1549L
Entrez Gene ID
25758
Gene Name
KIAA1549 like
Gene Aliases
C11orf41, C11orf69, G2
Chromosome Location
Chr. 11: 33376569 - 33674102 on Build GRCh38
UniGene ID
Hs.502266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012194.2 NP_036326.2 20 10987
XM_017017484.1 XP_016872973.1 20 11005
GenBank mRNA AK094307.1 - 1 1743
BC025668.1 - 1 1037

View Full Product Details