Gene Transcripts

Gene Symbol
C22orf31
Entrez Gene ID
25770
Gene Name
chromosome 22 open reading frame 31
Gene Aliases
HS747E2A, bK747E2.1
Chromosome Location
Chr. 22: 29058672 - 29073974 on Build GRCh38
UniGene ID
Hs.50891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015370.1 NP_056185.1 2 409
XM_011530096.2 XP_011528398.1 2 377
XM_017028741.1 XP_016884230.1 2 332
GenBank mRNA AL035364.1 - 2 409
BC074758.2 - 2 327 30915217
BC126262.1 - 2 331 8991978
BC126264.1 - 2 331 8991980
CR456405.1 - 2 296

View Full Product Details